3-Benzyloxy-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Benzyloxy)-4-methoxyphenylboronic acid, (3-(Benzyloxy)-4-methoxyphenyl)boronic acid

29161-1G 29161-25G 29161-500MG 29161-5G
76913-174EA 213.25 USD
76913-174 76913-176 76913-178 76913-180
3-Benzyloxy-4-methoxyphenylboronic acid ≥97%
3-Benzyloxy-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₄
MW: 258.0775 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696652
CAS Number: 243990-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR