3-Hydroxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29100-25G 29100-5G
76913-162EA 534.7 USD
76913-162 76913-164
3-Hydroxy-2-methoxybenzaldehyde ≥97%
3-Hydroxy-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.147 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06656119
CAS Number: 66495-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR