3-Bromo-5-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

29072-25G 29072-5G
76913-134EA 219.61 USD
76913-134 76913-136
3-Bromo-5-methoxybenzonitrile ≥97%
3-Bromo-5-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD08061971
CAS Number: 867366-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR