4-(3-Bromophenyl)-tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

29004-1G 29004-25G 29004-500MG 29004-5G
76913-112EA 215.37 USD
76913-112 76913-114 76913-116 76913-118
4-(3-Bromophenyl)-tetrahydro-2H-pyran-4-ol ≥97%
4-(3-Bromophenyl)-tetrahydro-2H-pyran-4-ol
Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Refrigerator
CAS Number: 135048-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR