4-Bromo-2,6-dichloro-3-methylphenol ≥95%

Supplier: AOBChem USA

Warning

28889-1G 28889-500MG 28889-5G
76913-088EA 498.63 USD
76913-088 76913-090 76913-092
4-Bromo-2,6-dichloro-3-methylphenol ≥95%
4-Bromo-2,6-dichloro-3-methylphenol
Formula: C₇H₅BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD28973115
CAS Number: 56037-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR