3-Bromo-5-chloro-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

28788-1G 28788-25G 28788-5G
76913-064EA 96.55 USD
76913-064 76913-066 76913-068
3-Bromo-5-chloro-2-fluorobenzoic acid ≥97%
3-Bromo-5-chloro-2-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21607478
CAS Number: 1269232-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR