3-Hydroxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

28717-1G 28717-25G 28717-5G
76913-050EA 109.28 USD
76913-050 76913-052 76913-054
3-Hydroxy-2-methylbenzaldehyde ≥97%
3-Hydroxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD09999328
CAS Number: 90111-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR