3-Hydroxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

28703-25G 28703-5G
76913-040EA 453.01 USD
76913-040 76913-042
3-Hydroxy-5-methylbenzoic acid ≥97%
3-Hydroxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.147 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09833213
CAS Number: 585-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR