3-Bromo-5-ethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

28529-1G 28529-25G 28529-500MG 28529-5G
76913-018EA 350.1 USD
76913-018 76913-020 76913-022 76913-024
3-Bromo-5-ethoxyphenol ≥97%
3-Bromo-5-ethoxyphenol
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12405424
CAS Number: 1026796-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR