3,6-Difluorosalicylic acid ≥97%

Supplier: AOBChem USA

Warning

28407-1G 28407-25G 28407-5G
76912-996EA 102.91 USD
76912-996 76912-998 76913-000
3,6-Difluorosalicylic acid ≥97%
3,6-Difluorosalicylic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08275285
CAS Number: 749230-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR