3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3,5-Difluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

28366-1G 28366-25G 28366-5G
76912-988EA 94.43 USD
76912-988 76912-990 76912-992
3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester ≥97%
3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781207
CAS Number: 1073354-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR