4,5-Difluoro-2-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

28171-1G 28171-25G 28171-500MG 28171-5G
76912-956EA 265.23 USD
76912-956 76912-958 76912-960 76912-962
4,5-Difluoro-2-formylphenylboronic acid ≥97%
4,5-Difluoro-2-formylphenylboronic acid
Formula: C₇H₅BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD20529455
CAS Number: 1432610-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR