3-Bromo-5-(2-methoxyethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

28065-1G 28065-25G 28065-500MG 28065-5G
76912-942EA 129.43 USD
76912-942 76912-944 76912-946 76912-948
3-Bromo-5-(2-methoxyethoxy)pyridine ≥97%
3-Bromo-5-(2-methoxyethoxy)pyridine
Formula: C₈H₁₀BrNO₂
MW: 232.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19674031
CAS Number: 181585-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR