3-Benzyloxy-4-bromobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzyloxy)-1-bromo-4-(trifluoromethyl)benzene

27959-1G 27959-25G 27959-5G
76912-922EA 82.76 USD
76912-922 76912-924 76912-926
3-Benzyloxy-4-bromobenzotrifluoride ≥97%
3-Benzyloxy-4-bromobenzotrifluoride
Formula: C₁₄H₁₀BrF₃O
MW: 331.128 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142627
CAS Number: 1429027-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR