4-(Phenylcarbamoyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: [4-(Phenylamino-1-carbonyl)phenyl] boronic acid pinacol ester, 4-(Phenylaminocarbonyl)benzeneboronic acid pinacol ester

27918-25G 27918-5G
76912-914EA 190.97 USD
76912-914 76912-916
4-(Phenylcarbamoyl)phenylboronic acid pinacol ester ≥97%
4-(Phenylcarbamoyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD12546047
CAS Number: 949115-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR