3-(4-Nitrophenoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

27869-1G 27869-25G 27869-500MG 27869-5G
76912-900EA 126.25 USD
76912-900 76912-902 76912-904 76912-906
3-(4-Nitrophenoxy)pyridine ≥97%
3-(4-Nitrophenoxy)pyridine
Formula: C₁₁H₈N₂O₃
MW: 216.193 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05855249
CAS Number: 28232-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR