3-Bromo-5-fluoro-2-methoxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

27745-1G 27745-25G 27745-500MG 27745-5G
76912-858EA 142.17 USD
76912-858 76912-860 76912-862 76912-864
3-Bromo-5-fluoro-2-methoxybenzoic acid ≥97% research grade
3-Bromo-5-fluoro-2-methoxybenzoic acid
Formula: C₈H₆BrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28142617
CAS Number: 1254340-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR