3-Bromo-5-fluoro-4-hydroxypyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-fluoropyridin-4-ol

27546-1G 27546-25G 27546-500MG 27546-5G
76912-840EA 173.99 USD
76912-840 76912-842 76912-844 76912-846
3-Bromo-5-fluoro-4-hydroxypyridine ≥95%
3-Bromo-5-fluoro-4-hydroxypyridine
Formula: C₅H₃BrFNO
MW: 191.986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185370
CAS Number: 1214342-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR