3-Fluoro-4-(trimethylsilyl)pyridine-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

27494-25G 27494-5G
76912-828EA 283.27 USD
76912-828 76912-830
3-Fluoro-4-(trimethylsilyl)pyridine-2-carbaldehyde ≥97%
3-Fluoro-4-(trimethylsilyl)pyridine-2-carbaldehyde
Formula: C₉H₁₂FNOSi
MW: 197.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142594
CAS Number: 1809157-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR