3,6-Difluorosalicylaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3,6-Difluoro-2-hydroxybenzaldehyde

27407-1G 27407-25G 27407-5G
76912-802EA 93.36 USD
76912-802 76912-804 76912-806
3,6-Difluorosalicylaldehyde ≥95%
3,6-Difluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08275281
CAS Number: 502762-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR