3,6-Dibromo-2-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

27343-1G 27343-25G 27343-5G
76912-780EA 82.76 USD
76912-780 76912-782 76912-784
3,6-Dibromo-2-chloropyridine ≥97%
3,6-Dibromo-2-chloropyridine
Formula: C₅H₂Br₂ClN
Storage Temperature: Refrigerator
MDL Number: MFCD09037464
CAS Number: 942206-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR