3-Fluoro-5-pyrrolidin-1-ylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27299-1G 27299-25G 27299-500MG 27299-5G
76912-756EA 201.58 USD
76912-756 76912-758 76912-760 76912-762
3-Fluoro-5-pyrrolidin-1-ylbenzoic acid ≥97%
3-Fluoro-5-pyrrolidin-1-ylbenzoic acid
Formula: C₁₁H₁₂FNO₂
MW: 209.217 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22083307
CAS Number: 874133-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR