3-Hydroxy-2-methoxypyridine-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

27260-1G 27260-500MG 27260-5G
76912-718EA 736.27 USD
76912-718 76912-720 76912-722
3-Hydroxy-2-methoxypyridine-5-boronic acid pinacol ester ≥97%
3-Hydroxy-2-methoxypyridine-5-boronic acid pinacol ester
Formula: C₁₂H₁₈BNO₄
MW: 251.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26516759
CAS Number: 1857348-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR