3-(Propan-2-yloxy)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Isopropoxyphenol

27243-1G 27243-25G 27243-5G
76912-704EA 112.46 USD
76912-704 76912-706 76912-708
3-(Propan-2-yloxy)phenol ≥97%
3-(Propan-2-yloxy)phenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11103458
CAS Number: 68792-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR