3-(Benzyloxy)phenol ≥97%

Supplier: AOBChem USA

Warning

27225-25G 27225-5G
76912-692EA 339.49 USD
76912-692 76912-694
3-(Benzyloxy)phenol ≥97%
3-(Benzyloxy)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00134682
CAS Number: 3769-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR