3,5-Dibromo-2-hydroxy-6-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

27207-25G 27207-5G
76912-684EA 418 USD
76912-684 76912-686
3,5-Dibromo-2-hydroxy-6-methylpyridine ≥97% research grade
3,5-Dibromo-2-hydroxy-6-methylpyridine
Formula: C₆H₅Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD04066716
CAS Number: 500587-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR