3-Bromo-6-fluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27163-1G 27163-25G 27163-500MG 27163-5G
76912-670EA 350.1 USD
76912-670 76912-672 76912-674 76912-676
3-Bromo-6-fluoro-2-methoxybenzoic acid ≥97%
3-Bromo-6-fluoro-2-methoxybenzoic acid
Formula: C₈H₆BrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28015437
CAS Number: 1426073-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR