2-(1-Piperidinyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 3-Piperidinophenylboronic acid pinacol ester

27076-1G 27076-25G 27076-5G
76912-630EA 91.24 USD
76912-630 76912-632 76912-634
2-(1-Piperidinyl)phenylboronic acid pinacol ester ≥97% research grade
2-(1-Piperidinyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₂
MW: 287.2100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368528
CAS Number: 852227-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR