3-Cyano-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27015-1G 27015-25G 27015-5G
76912-616EA 109.28 USD
76912-616 76912-618 76912-620
3-Cyano-2-methylphenylboronic acid ≥97%
3-Cyano-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD18394417
CAS Number: 313546-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR