3-Bromo-2-methoxy-4-picoline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-methoxy-4-methylpyridine

26980-1G 26980-25G 26980-5G
76912-608EA 62.6 USD
76912-608 76912-610 76912-612
3-Bromo-2-methoxy-4-picoline ≥97%
3-Bromo-2-methoxy-4-picoline

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061305
CAS Number: 717843-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR