3-Fluoro-5-morpholino-1-bromobenzene ≥95%

Supplier: AOBChem USA

Warning

26919-25G 26919-5G
76912-580EA 595.17 USD
76912-580 76912-582
3-Fluoro-5-morpholino-1-bromobenzene ≥95%
3-Fluoro-5-morpholino-1-bromobenzene
Formula: C₁₀H₁₁BrFNO
MW: 260.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546522
CAS Number: 1129541-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR