4-(1-Piperidylmethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Piperidin-1-ylmethyl)phenylboronic acid

26758-1G 26758-25G 26758-500MG 26758-5G
76912-522EA 290.69 USD
76912-522 76912-524 76912-526 76912-528
4-(1-Piperidylmethyl)phenylboronic acid ≥95%
4-(1-Piperidylmethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06801706
CAS Number: 1200434-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR