3-Formyl-4-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Formyl-4-hydroxyphenyl)boronic acid

26645-1G 26645-25G 26645-500MG 26645-5G
76912-482EA 379.81 USD
76912-482 76912-484 76912-486 76912-488
3-Formyl-4-hydroxyphenylboronic acid ≥97%
3-Formyl-4-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BO₄
MW: 165.939 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13186921
CAS Number: 182344-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR