4-Bromo-2-chloro-5-methylphenol ≥97% research grade

Supplier: AOBChem USA

Warning

26578-1G 26578-25G 26578-500MG 26578-5G
76912-466EA 177.18 USD
76912-466 76912-468 76912-470 76912-472
4-Bromo-2-chloro-5-methylphenol ≥97% research grade
4-Bromo-2-chloro-5-methylphenol
Formula: C₇H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD18459215
CAS Number: 319473-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR