2-Methyl-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)propan-2-amine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Tert-butylaminomethyl)phenylboronic acid pinacol ester

26556-25G 26556-5G
76912-442EA 275.84 USD
76912-442 76912-444
2-Methyl-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)propan-2-amine ≥95%
2-Methyl-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)propan-2-amine
Formula: C₁₇H₂₈BNO₂
MW: 289.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546203
CAS Number: 1628014-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR