4-(Dimethylamino)-5-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26543-1G 26543-500MG 26543-5G
76912-428EA 587.74 USD
76912-428 76912-430 76912-432
4-(Dimethylamino)-5-formylphenylboronic acid pinacol ester ≥97%
4-(Dimethylamino)-5-formylphenylboronic acid pinacol ester
Formula: C₁₅H₂₂BNO₃
MW: 275.151 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22493682
CAS Number: 1356400-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR