3-Iodo-4-methoxypyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
26510-25G
26510-5G
76912-416EA
418
USD
76912-416
76912-418
3-Iodo-4-methoxypyridine ≥97%
3-Iodo-4-methoxypyridine
Formula:
C₆H₆INO Storage Temperature: Ambient |
MDL Number:
MFCD01416617 CAS Number: 89640-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...