3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Chloro-2-fluoropyridne-5-boronic acid pinacol ester

26473-1G 26473-25G 26473-5G
76912-396EA 96.55 USD
76912-396 76912-398 76912-400
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester ≥97%
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BClFNO₂
MW: 257.497 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16995268
CAS Number: 1220219-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR