3-Chloro-4-methylbenzyl alcohol ≥95% research grade

Supplier: AOBChem USA

Warning

26467-25G 26467-5G
76912-388EA 183.54 USD
76912-388 76912-390
3-Chloro-4-methylbenzyl alcohol ≥95% research grade
3-Chloro-4-methylbenzyl alcohol
Formula: C₈H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD00016863
CAS Number: 39652-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR