3-Bromo-2-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

26334-1G 26334-25G 26334-500MG 26334-5G
76912-324EA 118.83 USD
76912-324 76912-326 76912-328 76912-330
3-Bromo-2-methylbenzamide ≥95%
3-Bromo-2-methylbenzamide
Formula: C₈H₈BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD17170782
CAS Number: 919363-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR