3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxy-5-methylphenylboronic acid, pinacol ester

26226-1G 26226-25G 26226-500MG 26226-5G
76912-276EA 260.99 USD
76912-276 76912-278 76912-280 76912-282
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formula: C₁₃H₁₉BO₃
MW: 234.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12546605
CAS Number: 889659-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR