3-Formyl-4-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
26219-1G
26219-25G
26219-500MG
26219-5G
76912-260EA
292.81
USD
76912-260
76912-262
76912-264
76912-266
3-Formyl-4-methoxybenzoic acid ≥97%
3-Formyl-4-methoxybenzoic acid
Formula:
C₉H₈O₄ MW: 180.157 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12405165 CAS Number: 91420-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...