3-Bromo-5-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Brom-5-hydroxyanisole

26215-1G 26215-25G 26215-500MG 26215-5G
76912-246EA 168.69 USD
76912-246 76912-248 76912-250 76912-252
3-Bromo-5-methoxyphenol ≥97%
3-Bromo-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12405420
CAS Number: 855400-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR