4-(4-Trifluoromethoxyphenyl)phenol ≥97%
Supplier: AOBChem USA
|
Warning
|
26205-1G
26205-25G
26205-500MG
26205-5G
76912-230EA
132.62
USD
76912-230
76912-232
76912-234
76912-236
4-(4-Trifluoromethoxyphenyl)phenol ≥97%
4-(4-Trifluoromethoxyphenyl)phenol
Formula:
C₁₃H₉F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18312997 CAS Number: 173025-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...