4-Amino-3-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

26101-1G 26101-25G 26101-500MG 26101-5G
76912-170EA 298.12 USD
76912-170 76912-172 76912-174 76912-176
4-Amino-3-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97% research grade
4-Amino-3-(trifluoromethoxy)phenylboronic acid pinacol ester
Formula: C₁₃H₁₇BF₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16996388
CAS Number: 1256360-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR