3-Bromo-4-chloro-5-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

26090-1G 26090-25G 26090-500MG 26090-5G
76912-160EA 178.24 USD
76912-160 76912-162 76912-164 76912-166
3-Bromo-4-chloro-5-fluoropyridine ≥97%
3-Bromo-4-chloro-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
Storage Temperature: Refrigerator
MDL Number: MFCD18257571
CAS Number: 1211540-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR