1-Methyl-1H-imidazole-4,5-dicarboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4,5-Dicarboxy-1-methyl-1H-imidazole, 1-Methylimidazole-4,5-dicarboxylic Acid
25972-25G
25972-5G
76912-132EA
204.76
USD
76912-132
76912-134
1-Methyl-1H-imidazole-4,5-dicarboxylic acid ≥97%
1-Methyl-1H-imidazole-4,5-dicarboxylic acid
Formula:
C₆H₆N₂O₄ MW: 170.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01552045 CAS Number: 19485-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...