3-Bromo-2-fluoro-6-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

25942-1G 25942-25G 25942-500MG 25942-5G
76912-118EA 148.53 USD
76912-118 76912-120 76912-122 76912-124
3-Bromo-2-fluoro-6-methoxybenzoic acid ≥95%
3-Bromo-2-fluoro-6-methoxybenzoic acid
Formula: C₈H₆BrFO₃
MW: 249.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24848707
CAS Number: 1449008-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR