(3-Fluoro-2-pyridinyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-2-(hydroxymethyl)pyridine, (3-Fluoropyrid-2-yl)methanol

25916-25G 25916-5G
76912-110EA 275.84 USD
76912-110 76912-112
(3-Fluoro-2-pyridinyl)methanol ≥97%
(3-Fluoro-2-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.1200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09025826
CAS Number: 31181-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR