3-Bromo-2-fluoro-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25819-1G 25819-25G 25819-500MG 25819-5G
76912-062EA 153.84 USD
76912-062 76912-064 76912-066 76912-068
3-Bromo-2-fluoro-5-(trifluoromethyl)benzaldehyde ≥97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O
MW: 271.006 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23713159
CAS Number: 1236538-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR